3-(Trifluormethyl)-N-tert-butylbenzamid

Lieferant: COMBI-BLOCKS

CA-4987-5G CA-4987-1G
COBBCA-4987-5GEA 127 EUR
COBBCA-4987-5G COBBCA-4987-1G
3-(Trifluormethyl)-N-tert-butylbenzamid
3-(Trifluormethyl)-N-tert-butylbenzamid
Formel: C₁₂H₁₄F₃NO
MDL: MFCD16618941
CAS-Nummer: 340136-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR