4-(Trifluormethyl)-N-tert-butylbenzamid

Lieferant: COMBI-BLOCKS

CA-4986-5G CA-4986-1G
COBBCA-4986-5GEA 595 EUR
COBBCA-4986-5G COBBCA-4986-1G
4-(Trifluormethyl)-N-tert-butylbenzamid
4-(Trifluormethyl)-N-tert-butylbenzamid
Formel: C₁₂H₁₄F₃NO
MDL: MFCD16618940
CAS-Nummer: 91888-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR