Methyl-4-iod-3-methylbenzoat ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 4-iodo-3-methylbenzoate

B25395.22 B25395.14 B25395.06
B25395.22EA 502 EUR
B25395.22 B25395.14 B25395.06
Methyl-4-iod-3-methylbenzoat ≥98%
Methyl-4-iod-3-methylbenzoat

Purity: 98% CAS No.: 5471-81-8 Molecular Formula: C9H9IO2 Formula Weight: 276.07 Melting Point: 59-62° MDL No.: MFCD02683863

Formel: C₉H₉IO₂
Molecular Weight: 276,07 g/mol
Schmelzpunkt: 59…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02683863
CAS-Nummer: 5471-81-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR