3,4-Dimethoxybenzamidin Hydrochlorid
Lieferant: CHEMPUR
Synonyme:
3,4-Dimethoxybenzimidamide hydrochloride
12-0023.1
12-0023.5
CHMP12-0023.1EA
0
EUR
CHMP12-0023.1
CHMP12-0023.5
3,4-Dimethoxybenzamidin Hydrochlorid
3,4-Dimethoxybenzamidin Hydrochlorid
Formel:
C₉H₁₃ClN₂O₂ |
MDL:
MFCD01667882 CAS-Nummer: 51488-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...