2,3,4-Trimethoxybenzamidin Hydrochlorid

Lieferant: CHEMPUR

CHMP12-0039.1EA 0 EUR
CHMP12-0039.1 CHMP12-0039.5
2,3,4-Trimethoxybenzamidin Hydrochlorid
2,3,4-Trimethoxybenzamidin Hydrochlorid
Formel: C₁₀H₁₅ClN₂O₃
MDL: MFCD04114424
CAS-Nummer: 1187929-17-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR