2,4,6-Trimethoxybenzamidin Hydrochlorid

Lieferant: CHEMPUR

12-0040.1 12-0040.5
CHMP12-0040.1EA 0 EUR
CHMP12-0040.1 CHMP12-0040.5
2,4,6-Trimethoxybenzamidin Hydrochlorid
2,4,6-Trimethoxybenzamidin Hydrochlorid
Formel: C₁₀H₁₅ClN₂O₃
MDL: MFCD04114425
CAS-Nummer: 1187929-24-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR