3,4,5-Trimethoxybenzamidin Hydrochlorid

Lieferant: CHEMPUR

12-0033.1 12-0033.5
CHMP12-0033.1EA 0 EUR
CHMP12-0033.1 CHMP12-0033.5
3,4,5-Trimethoxybenzamidin Hydrochlorid
3,4,5-Trimethoxybenzamidin Hydrochlorid
Formel: C₁₀H₁₅ClN₂O₃
MDL: MFCD07186251
CAS-Nummer: 4156-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR