N-(2,2-Dimethoxyethyl)-4-methylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0839-5G OT-0839-1G
COBBOT-0839-5GEA 0 EUR
COBBOT-0839-5G COBBOT-0839-1G
N-(2,2-Dimethoxyethyl)-4-methylbenzolsulfonamid
N-(2,2-Dimethoxyethyl)-4-methylbenzolsulfonamid
Formel: C₁₁H₁₇NO₄S
MDL: MFCD00971347
CAS-Nummer: 58754-95-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR