N-Benzyl-3-brom-4-methylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0159-5G OT-0159-1G
COBBOT-0159-5GEA 595 EUR
COBBOT-0159-5G COBBOT-0159-1G
N-Benzyl-3-brom-4-methylbenzolsulfonamid
N-Benzyl-3-brom-4-methylbenzolsulfonamid
Formel: C₁₄H₁₄BrNO₂S
MDL: MFCD07363832
CAS-Nummer: 850429-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR