Methyl-3,5-bis(trifluormethyl)benzoat

Lieferant: FLUOROCHEM

Synonyme: Methyl 3,5-bis(trifluoromethyl)benzoate

006796-5G 006796-25G
FLUO006796-5GEA 0 EUR
FLUO006796-5G FLUO006796-25G
Methyl-3,5-bis(trifluormethyl)benzoat
Methyl-3,5-bis(trifluormethyl)benzoat
Formel: C₁₀H₆F₆O₂
MDL: MFCD00052317
CAS-Nummer: 26107-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR