3-Brom-5-chlor-2-(4-morpholino)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 4-(3-Bromo-5-chloropyridin-2-yl)morpholine

PY-1244-1G PY-1244-5G
COBBPY-1244-1GEA 204 EUR
COBBPY-1244-1G COBBPY-1244-5G
3-Brom-5-chlor-2-(4-morpholino)pyridin
3-Brom-5-chlor-2-(4-morpholino)pyridin
Formel: C₉H₁₀BrClN₂O
Molecular Weight: 277.546 g/mol
MDL: MFCD13195782
CAS-Nummer: 1199773-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR