5-Brom-3-chlor-2-(4-morpholino)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 4-(5-Bromo-3-chloropyridin-2-yl)morpholine

PY-1219-1G PY-1219-5G PY-1219-250M
COBBPY-1219-1GEA 310 EUR
COBBPY-1219-1G COBBPY-1219-5G COBBPY-1219-250M
5-Brom-3-chlor-2-(4-morpholino)pyridin
5-Brom-3-chlor-2-(4-morpholino)pyridin
Formel: C₉H₁₀BrClN₂O
Molecular Weight: 277.5500 g/mol
MDL: MFCD13195777
CAS-Nummer: 1199773-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR