3-Cyan-2-(4-morpholino)pyridin

Lieferant: BIONET RESEARCH

Synonyme: 2-(Morpholin-4-yl)nicotinonitrile

3X-0726.500 3X-0726.1 3X-0726.5 3X-0726.10
BIOT3X-0726.500EA 0 EUR
BIOT3X-0726.500 BIOT3X-0726.1 BIOT3X-0726.5 BIOT3X-0726.10
3-Cyan-2-(4-morpholino)pyridin
3-Cyan-2-(4-morpholino)pyridin
Formel: C₁₀H₁₁N₃O
MDL: MFCD04113612
CAS-Nummer: 59025-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR