3-Cyan-2-(4-morpholino)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Morpholin-4-yl)nicotinonitrile

PY-8043-5G PY-8043-1G
COBBPY-8043-5GEA 402 EUR
COBBPY-8043-5G COBBPY-8043-1G
3-Cyan-2-(4-morpholino)pyridin
3-Cyan-2-(4-morpholino)pyridin
Formel: C₁₀H₁₁N₃O
MDL: MFCD04113612
CAS-Nummer: 59025-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR