2-(4-Morpholino)-3-nitropyridin

Lieferant: BIONET RESEARCH

Synonyme: 4-(3-Nitro-2-pyridinyl)morpholine, 4-(3-Nitropyridin-2-yl)morpholine

8R-0394.500 8R-0394.1 8R-0394.5 8R-0394.10
BIOT8R-0394.500EA 0 EUR
BIOT8R-0394.500 BIOT8R-0394.1 BIOT8R-0394.5 BIOT8R-0394.10
2-(4-Morpholino)-3-nitropyridin
2-(4-Morpholino)-3-nitropyridin
Formel: C₉H₁₁N₃O₃
Molecular Weight: 209.2019 g/mol
MDL: MFCD00452814
CAS-Nummer: 24255-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR