2-(4-Morpholino)-5-nitropyridin

Lieferant: BIONET RESEARCH

Synonyme: 4-(5-Nitropyridin-2-yl)morpholine

9R-0322.500 9R-0322.1 9R-0322.5 9R-0322.10
BIOT9R-0322.500EA 0 EUR
BIOT9R-0322.500 BIOT9R-0322.1 BIOT9R-0322.5 BIOT9R-0322.10
2-(4-Morpholino)-5-nitropyridin
2-(4-Morpholino)-5-nitropyridin
Formel: C₉H₁₁N₃O₃
Molecular Weight: 209.2019 g/mol
MDL: MFCD00023308
CAS-Nummer: 26820-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR