4-Cyclohexylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Cyclohexylbenzeneboronic acid, (4-Cyclohexylphenyl)boronic acid
BB-2905-5G
BB-2905-1G
COBBBB-2905-5GEA
862
EUR
COBBBB-2905-5G
COBBBB-2905-1G
4-Cyclohexylphenylboronsäure
4-Cyclohexylphenylboronsäure
Formel:
C₁₂H₁₇BO₂ Molecular Weight: 204.08 g/mol |
MDL:
MFCD02093068 CAS-Nummer: 374538-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...