2-(Cyclopropylmethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-9035-5G BB-9035-1G
COBBBB-9035-5GEA 0 EUR
COBBBB-9035-5G COBBBB-9035-1G
2-(Cyclopropylmethoxy)phenylboronsäure
2-(Cyclopropylmethoxy)phenylboronsäure
Formel: C₁₀H₁₃BO₃
MDL: MFCD04039015
CAS-Nummer: 1050510-36-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR