3-(Cyclopropylmethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(Cyclopropylmethoxy)phenyl)boronic acid

BB-9037-1G BB-9037-5G
COBBBB-9037-1GEA 303 EUR
COBBBB-9037-1G COBBBB-9037-5G
3-(Cyclopropylmethoxy)phenylboronsäure
3-(Cyclopropylmethoxy)phenylboronsäure
Formel: C₁₀H₁₃BO₃
Molecular Weight: 192,200 g/mol
MDL: MFCD04039016
CAS-Nummer: 411229-76-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR