3-(Benzyloxy)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-[3-(Benzyloxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(3-(Benzyloxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Benzyloxyphenylboronic acid, pinacol ester
PN-2153-5G
PN-2153-1G
COBBPN-2153-5GEA
0
EUR
COBBPN-2153-5G
COBBPN-2153-1G
3-(Benzyloxy)phenylboronsäurepinakolester
3-(Benzyloxy)phenylboronsäurepinakolester
Formel:
C₁₉H₂₃BO₃ |
MDL:
MFCD05663839 CAS-Nummer: 765908-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...