4-(Benzyloxy)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4-Benzyloxybenzeneboronic acid pinacol ester, 2-(4-(Benzyloxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-2151-5G
PN-2151-25G
PN-2151-1G
COBBPN-2151-5GEA
117
EUR
COBBPN-2151-5G
COBBPN-2151-25G
COBBPN-2151-1G
4-(Benzyloxy)phenylboronsäurepinakolester
4-(Benzyloxy)phenylboronsäurepinakolester
Formel:
C₁₉H₂₃BO₃ Molecular Weight: 310,19 g/mol |
MDL:
MFCD04974107 CAS-Nummer: 754226-40-9 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...