4-(Benzyl(ethyl)carbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3210-5G BB-3210-1G
COBBBB-3210-5GEA 0 EUR
COBBBB-3210-5G COBBBB-3210-1G
4-(Benzyl(ethyl)carbamoyl)phenylboronsäure
4-(Benzyl(ethyl)carbamoyl)phenylboronsäure
Formel: C₁₆H₁₈BNO₃
MDL: MFCD09027215
CAS-Nummer: 913835-41-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR