4-(Benzylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(Benzylcarbamoyl)phenyl)boronic acid, 4-(Benzylcarbamoyl)benzeneboronic acid, 4-(N-Benzylaminocarbonyl)phenylboronic acid

BB-3005-5G BB-3005-1G
COBBBB-3005-5GEA 0 EUR
COBBBB-3005-5G COBBBB-3005-1G
4-(Benzylcarbamoyl)phenylboronsäure
4-(Benzylcarbamoyl)phenylboronsäure
Formel: C₁₄H₁₄BNO₃
MDL: MFCD03411953
CAS-Nummer: 252663-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR