2-(Benzyloxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Benzyloxyphenylboronic acid, (2-(Benzyloxy)phenyl)boronic acid, 2-Benzyloxybenzeneboronic acid
BB-2466-5G
BB-2466-1G
BB-2466-25G
COBBBB-2466-5GEA
70.8
EUR
COBBBB-2466-5G
COBBBB-2466-1G
COBBBB-2466-25G
2-(Benzyloxy)phenylboronsäure
2-(Benzyloxy)phenylboronsäure
Formel:
C₁₃H₁₃BO₃ Molecular Weight: 228,06 g/mol |
MDL:
MFCD01632206 CAS-Nummer: 190661-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...