3-(Benzyloxy)phenylboronsäure
Lieferant: Thermo Scientific
Synonyme:
(3-(Benzyloxy)phenyl)boronic acid, 3-Benzyloxybenzeneboronic acid
AC35886.250
AC35886.1
MAYBAC35886.250EA
0
EUR
MAYBAC35886.250
MAYBAC35886.1
3-(Benzyloxy)phenylboronsäure
3-(Benzyloxy)phenylboronsäure
Formel:
C₁₃H₁₃BO₃ Molecular Weight: 228,06 g/mol |
MDL:
MFCD02093063 CAS-Nummer: 156682-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...