3-(Benzyloxy)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (3-(Benzyloxy)phenyl)boronic acid, 3-Benzyloxybenzeneboronic acid

M-Clarity™ Qualitätsstufe = MQ100

526339-25G 526339-5G
SIAL526339-25GEA 128 EUR
SIAL526339-25G SIAL526339-5G
3-(Benzyloxy)phenylboronsäure, Sigma-Aldrich®
3-(Benzyloxy)phenylboronsäure
Formel: C₁₃H₁₃BO₃
Molecular Weight: 228,06 g/mol
MDL: MFCD02093063
CAS-Nummer: 156682-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR