4-(Benzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Benzyloxybenzeneboronic acid, (4-(Benzyloxy)phenyl)boronic acid

BB-2151-5G BB-2151-25G BB-2151-100G
COBBBB-2151-5GEA 26 EUR
COBBBB-2151-5G COBBBB-2151-25G COBBBB-2151-100G
4-(Benzyloxy)phenylboronsäure
4-(Benzyloxy)phenylboronsäure
Formel: C₁₃H₁₃BO₃
Molecular Weight: 228,06 g/mol
MDL: MFCD01075705
CAS-Nummer: 146631-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR