3-(3'-Methoxybenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(3-Methoxybenzyloxy)phenylboronic acid

BB-8068-1G BB-8068-250M
COBBBB-8068-1GEA 0 EUR
COBBBB-8068-1G COBBBB-8068-250M
3-(3'-Methoxybenzyloxy)phenylboronsäure
3-(3'-Methoxybenzyloxy)phenylboronsäure
Formel: C₁₄H₁₅BO₄
Molecular Weight: 258.077 g/mol
MDL: MFCD07783019
CAS-Nummer: 871125-75-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR