3-Brom-4-[(3,4-dichlorbenzyl)oxy]-5-methoxybenzaldehyd

Lieferant: Thermo Scientific

BTB06313.250 BTB06313.1
MAYBBTB06313.250EA 0 EUR
MAYBBTB06313.250 MAYBBTB06313.1
3-Brom-4-[(3,4-dichlorbenzyl)oxy]-5-methoxybenzaldehyd
3-Brom-4-[(3,4-dichlorbenzyl)oxy]-5-methoxybenzaldehyd
Formel: C₁₅H₁₁BrCl₂O₃
CAS-Nummer: 219765-92-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR