2-(4-Brombenzamido)ethyl-4-brombenzoat

Lieferant: COMBI-BLOCKS

CA-4505-5G CA-4505-1G
COBBCA-4505-5GEA 1220 EUR
COBBCA-4505-5G COBBCA-4505-1G
2-(4-Brombenzamido)ethyl-4-brombenzoat
2-(4-Brombenzamido)ethyl-4-brombenzoat
Formel: C₁₆H₁₃Br₂NO₃
MDL: MFCD09878361
CAS-Nummer: 951885-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR