tert-Butyl-3-(benzyloxy)-4-brombenzoat

Lieferant: COMBI-BLOCKS

CA-4244-1G CA-4244-5G
COBBCA-4244-1GEA 98.3 EUR
COBBCA-4244-1G COBBCA-4244-5G
tert-Butyl-3-(benzyloxy)-4-brombenzoat
tert-Butyl-3-(benzyloxy)-4-brombenzoat
Formel: C₁₈H₁₉BrO₃
MDL: MFCD09027266
CAS-Nummer: 247186-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR