3-Benzyloxy-6-brom-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Benzyloxy)-6-bromo-2-fluorophenylboronic acid

BB-3982-5G BB-3982-1G
COBBBB-3982-5GEA 260 EUR
COBBBB-3982-5G COBBBB-3982-1G
3-Benzyloxy-6-brom-2-fluorphenylboronsäure
3-Benzyloxy-6-brom-2-fluorphenylboronsäure
Formel: C₁₃H₁₁BBrFO₃
Molecular Weight: 324.9400 g/mol
MDL: MFCD09475868
CAS-Nummer: 957035-10-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR