N-Benzyl-2-brom-5-(trifluormethoxy)benzolsulfonamid
Lieferant: COMBI-BLOCKS
HC-6244-5G
HC-6244-1G
COBBHC-6244-5GEA
0
EUR
COBBHC-6244-5G
COBBHC-6244-1G
N-Benzyl-2-brom-5-(trifluormethoxy)benzolsulfonamid
N-Benzyl-2-brom-5-(trifluormethoxy)benzolsulfonamid
Formel:
C₁₄H₁₁BrF₃NO₃S |
MDL:
MFCD09800993 CAS-Nummer: 951885-49-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...