2-Brom-N-(4-fluorbenzyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

HC-6219-5G HC-6219-1G
COBBHC-6219-5GEA 0 EUR
COBBHC-6219-5G COBBHC-6219-1G
2-Brom-N-(4-fluorbenzyl)benzolsulfonamid
2-Brom-N-(4-fluorbenzyl)benzolsulfonamid
Formel: C₁₃H₁₁BrFNO₂S
MDL: MFCD02556038
CAS-Nummer: 321705-40-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR