4-Brom-N-(4-methoxybenzyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0412-1G OT-0412-5G
COBBOT-0412-1GEA 292 EUR
COBBOT-0412-1G COBBOT-0412-5G
4-Brom-N-(4-methoxybenzyl)benzolsulfonamid
4-Brom-N-(4-methoxybenzyl)benzolsulfonamid
Formel: C₁₄H₁₄BrNO₃S
Molecular Weight: 356,2349 g/mol
MDL: MFCD01359619
CAS-Nummer: 329939-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR