N-Benzyl-2-brombenzolsulfonamid

Lieferant: COMBI-BLOCKS

HC-6217-5G HC-6217-1G
COBBHC-6217-5GEA 451 EUR
COBBHC-6217-5G COBBHC-6217-1G
N-Benzyl-2-brombenzolsulfonamid
N-Benzyl-2-brombenzolsulfonamid
Formel: C₁₃H₁₂BrNO₂S
MDL: MFCD02555935
CAS-Nummer: 321704-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR