N-Benzyl-3-brombenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0143-5G OT-0143-1G
COBBOT-0143-5GEA 303 EUR
COBBOT-0143-5G COBBOT-0143-1G
N-Benzyl-3-brombenzolsulfonamid
N-Benzyl-3-brombenzolsulfonamid
Formel: C₁₃H₁₂BrNO₂S
MDL: MFCD07363821
CAS-Nummer: 625470-36-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR