N-Benzyl-4-brombenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0081-5G OT-0081-25G OT-0081-1G OT-0081-100G
COBBOT-0081-5GEA 98.3 EUR
COBBOT-0081-5G COBBOT-0081-25G COBBOT-0081-1G COBBOT-0081-100G
N-Benzyl-4-brombenzolsulfonamid
N-Benzyl-4-brombenzolsulfonamid
Formel: C₁₃H₁₂BrNO₂S
Molecular Weight: 326,21 g/mol
MDL: MFCD00450587
CAS-Nummer: 3609-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR