3-Carbamoyl-4-chlorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2811-5G BB-2811-1G
COBBBB-2811-5GEA 0 EUR
COBBBB-2811-5G COBBBB-2811-1G
3-Carbamoyl-4-chlorphenylboronsäure
3-Carbamoyl-4-chlorphenylboronsäure
Formel: C₇H₇BClNO₃
MDL: MFCD07363796
CAS-Nummer: 871332-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR