4-Carbamoyl-2-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-Carbamoyl-2-fluorophenyl)boronic acid
BB-3318-5G
BB-3318-1G
COBBBB-3318-5GEA
1490
EUR
COBBBB-3318-5G
COBBBB-3318-1G
4-Carbamoyl-2-fluorphenylboronsäure
4-Carbamoyl-2-fluorphenylboronsäure
Formel:
C₇H₇BFNO₃ Molecular Weight: 182.9500 g/mol |
MDL:
MFCD09800867 CAS-Nummer: 874289-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...