2-(3'-Fluorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8006-1G BB-8006-250M
COBBBB-8006-1GEA 151 EUR
COBBBB-8006-1G COBBBB-8006-250M
2-(3'-Fluorbenzyloxy)phenylboronsäure
2-(3'-Fluorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₂BFO₃
MDL: MFCD07784392
CAS-Nummer: 871126-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR