2-(4'-Fluorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-((4-Fluorobenzyl)oxy)phenyl)boronic acid

BB-8014-1G BB-8014-250M
COBBBB-8014-1GEA 0 EUR
COBBBB-8014-1G COBBBB-8014-250M
2-(4'-Fluorbenzyloxy)phenylboronsäure
2-(4'-Fluorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₂BFO₃
MDL: MFCD07369749
CAS-Nummer: 870779-01-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR