4-Benzyloxy-2,3-difluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(Benzyloxy)-2,3-difluorophenyl)boronic acid, 4-(Benzyloxy)-2,3-difluorophenylboronic acid

BB-3815-1G BB-3815-5G
COBBBB-3815-1GEA 153 EUR
COBBBB-3815-1G COBBBB-3815-5G
4-Benzyloxy-2,3-difluorphenylboronsäure
4-Benzyloxy-2,3-difluorphenylboronsäure
Formel: C₁₃H₁₁BF₂O₃
Molecular Weight: 264.0325 g/mol
MDL: MFCD07644469
CAS-Nummer: 156635-87-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR