3-Benzyloxy-2,6-difluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3816-5G BB-3816-25G BB-3816-1G
COBBBB-3816-5GEA 273 EUR
COBBBB-3816-5G COBBBB-3816-25G COBBBB-3816-1G
3-Benzyloxy-2,6-difluorphenylboronsäure
3-Benzyloxy-2,6-difluorphenylboronsäure
Formel: C₁₃H₁₁BF₂O₃
Molecular Weight: 264,0325 g/mol
MDL: MFCD06200705
CAS-Nummer: 870718-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR