4-Benzyloxy-2-fluorphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 4-Benzyloxy-2-fluorobenzeneboronic acid, (4-(Benzyloxy)-2-fluorophenyl)boronic acid

016103-1G 016103-5G
FLUO016103-1GEA 109 EUR
FLUO016103-1G FLUO016103-5G
4-Benzyloxy-2-fluorphenylboronsäure
4-Benzyloxy-2-fluorphenylboronsäure
Formel: C₁₃H₁₂BFO₃
Molecular Weight: 246.04 g/mol
MDL: MFCD02093064
CAS-Nummer: 166744-78-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR