2-Benzyloxy-6-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-(Benzyloxy)-6-fluorophenyl)boronic acid
BB-3229-1G
BB-3229-5G
COBBBB-3229-1GEA
303
EUR
COBBBB-3229-1G
COBBBB-3229-5G
2-Benzyloxy-6-fluorphenylboronsäure
2-Benzyloxy-6-fluorphenylboronsäure
Formel:
C₁₃H₁₂BFO₃ Molecular Weight: 246.0420 g/mol |
MDL:
MFCD11617256 CAS-Nummer: 1217500-53-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...