3-(2'-Fluorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8036-1G BB-8036-250M
COBBBB-8036-1GEA 102 EUR
COBBBB-8036-1G COBBBB-8036-250M
3-(2'-Fluorbenzyloxy)phenylboronsäure
3-(2'-Fluorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₂BFO₃
MDL: MFCD07369750
CAS-Nummer: 849062-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR