3-(3'-Fluorbenzyloxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-((3-Fluorobenzyl)oxy)phenyl)boronic acid
BB-8077-1G
BB-8077-250M
COBBBB-8077-1GEA
330
EUR
COBBBB-8077-1G
COBBBB-8077-250M
3-(3'-Fluorbenzyloxy)phenylboronsäure
3-(3'-Fluorbenzyloxy)phenylboronsäure
Formel:
C₁₃H₁₂BFO₃ Molecular Weight: 246,0420 g/mol |
MDL:
MFCD08276822 CAS-Nummer: 1072951-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...