3-Benzyloxy-5-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Benzyloxy-5-fluorobenzeneboronic acid

BB-2778-5G BB-2778-1G
COBBBB-2778-5GEA 379 EUR
COBBBB-2778-5G COBBBB-2778-1G
3-Benzyloxy-5-fluorphenylboronsäure
3-Benzyloxy-5-fluorphenylboronsäure
Formel: C₁₃H₁₂BFO₃
Molecular Weight: 246.04 g/mol
MDL: MFCD07363782
CAS-Nummer: 850589-56-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR