2-Benzyloxy-5-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Benzyloxy)-5-fluorophenyl)boronic acid, 2-Benzyloxy-5-fluorobenzeneboronic acid

BB-2681-1G BB-2681-5G
COBBBB-2681-1GEA 49.7 EUR
COBBBB-2681-1G COBBBB-2681-5G
2-Benzyloxy-5-fluorphenylboronsäure
2-Benzyloxy-5-fluorphenylboronsäure
Formel: C₁₃H₁₂BFO₃
Molecular Weight: 246.0420 g/mol
MDL: MFCD03788403
CAS-Nummer: 779331-47-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR